Reagents Ammonium Bifluoride, Purified, Fiber Drum, 25 kg

Representative image only.

Reagents Ammonium Bifluoride, Purified, Fiber Drum, 25 kg

Reagents | Mfr: C2130500-25F4| Item: EW-62283-84

Manufactured using the highest quality raw materials in ISO 9001 facility

$1,435.00USD / Each

$1,435.00USD / Each
Product Description

Key Features

  • Colorless/white solid
  • Density: 1.5
  • Boiling point: approximately 240 °C (464 °F)
  • Solubility information: 630 g/L at 20 °C (68 °F)
  • 25 kg fiber drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaNH4F·HF
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • SynonymsAmmonium hydrogen fluoride
  • Weight (g)25000
  • Formula Weight57.04
  • Vapor Pressure (torr)0.7501
  • PackagingDrum
  • DOT InformationAMMONIUM HYDROGENDIFLUORIDE, SOLID, 8, P.G. II
  • Storage SymbolAmbient
  • CAS Number1341-49-7
  • UN NumberUN1727
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)124.6
  • Melting Point F (° F)256.28
  • DescriptionAmmonium Bifluoride, Purified, Fiber Drum, 25 kg
Documents & Certificates
Reviews
Q&A