Reagents Ammonium Carbonate, 500g, ACS Reagent

Representative image only.

Reagents Ammonium Carbonate, 500g, ACS Reagent

Reagents | Mfr: C1131500-500A5| Item: EW-78910-21

Manufactured using the highest quality raw materials in ISO 9001 facility

$235.75USD / Each

$235.75USD / Each
Product Description

Key Features

  • Quality and consistency your lab can count on
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

5 Year Warranty
Specifications
  • Purity GradeACS
  • Linear Formula(NH4)2CO3
  • ClassSalts
  • FormSolid
  • Weight (g)2500
  • Formula Weight96.09
  • PackagingPoly Jar
  • CAS Number8000-73-5
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionAmmonium Carbonate, 500g, ACS Reagent
  • Warranty5 years
Documents & Certificates
Reviews
Q&A