Reagents Ammonium Hydroxide, 2% (v/v), Volumetric, Natural Poly Bottle, 4 L

Representative image only.

Reagents Ammonium Hydroxide, 2% (v/v), Volumetric, Natural Poly Bottle, 4 L

Reagents | Mfr: CA011700-4A| Item: EW-78920-71

Manufactured using the highest quality raw materials in ISO 9001 facility

$145.50USD / Each

$145.50USD / Each
Product Description

Key Features

  • Aqueous
  • Colorless liquid
  • Density: 1
  • Solubility information: miscible
  • pH range: 11.4
  • 4 L natural poly bottle
  • 2 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Two year warranty
Specifications
  • Concentration2% v/v
  • ClassVolumetric
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)4000
  • PackagingPoly bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • Shelf Life (days)730
  • Shelf Life (months)24
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • DescriptionAmmonium Hydroxide, 2% (v/v), Volumetric, Natural Poly Bottle, 4 L
  • Warranty2 year
Documents & Certificates
Reviews
Q&A