Reagents Ammonium Hydroxide, ACS Reagent, Winpak Container, 20 L

Representative image only.

Reagents Ammonium Hydroxide, ACS Reagent, Winpak Container, 20 L

Reagents | Mfr: C5103500-20E7| Item: EW-78927-02

Manufactured using the highest quality raw materials in ISO 9001 facility

$528.00USD / Each

$528.00USD / Each
Product Description

Key Features

  • Assay 28.0 to 30.0%
  • Colorless liquid
  • Density: 0.9
  • Solubility information: miscible
  • pH range: 12.2
  • 20 L Winpak container
  • 3 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

3 Year Warranty
Specifications
  • Purity GradeACS
  • Linear FormulaNH4OH
  • ClassSalts
  • FormLiquid
  • Volume (mL)20000
  • Formula Weight35.05
  • PackagingWinpak container
  • DOT InformationAMMONIA SOLUTIONS, 8, P.G. III
  • Storage SymbolAmbient
  • CAS Number1336-21-6
  • UN NumberUN2672
  • Shelf Life (days)1125
  • Shelf Life (months)36
  • Boiling Point C (° C)24.7
  • Boiling Point F (° F)76.46
  • Melting Point C (° C)-91.5
  • Melting Point F (° F)-132.7
  • DescriptionAmmonium Hydroxide, ACS Reagent, Winpak Container, 20 L
  • Warranty3 year
Documents & Certificates
Reviews
Q&A