Reagents Ammonium Oxalate, Purified, Wide-Mouth Amber Glass Bottle, 500 g

Representative image only.

Reagents Ammonium Oxalate, Purified, Wide-Mouth Amber Glass Bottle, 500 g

Reagents | Mfr: C2136000-500C2| Item: EW-78918-54

Manufactured using the highest quality raw materials in ISO 9001 facility

$139.50USD / Each

$139.50USD / Each
Product Description

Key Features

  • Colorless/white solid
  • Density: 1.5
  • Solubility information: 45 g/L at 20 °C (68 °F)
  • 500 g wide-mouth amber glass bottle
  • 20 years warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

20 Year Warranty
Specifications
  • Purity GradePurified
  • Linear Formula(COONH4)2·H2O
  • ClassSalts
  • FormSolid
  • SynonymsAmmonium oxalate monohydrate
  • Weight (g)500
  • Formula Weight142.11
  • PackagingWide-mouth amber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number6009-70-7
  • Shelf Life (days)7325
  • Shelf Life (months)240
  • Melting Point C (° C)70
  • Melting Point F (° F)158
  • DescriptionAmmonium Oxalate, Purified, Wide-Mouth Amber Glass Bottle, 500 g
  • Warranty20 years
Documents & Certificates
Reviews
Q&A