Reagents Barium Chloride, 25% (w/v) with Surfactant, Volumetric, Natural Poly Bottle, 4 L

Representative image only.

Reagents Barium Chloride, 25% (w/v) with Surfactant, Volumetric, Natural Poly Bottle, 4 L

Reagents | Mfr: C7147010-4A| Item: EW-78922-70

Manufactured using the highest quality raw materials in ISO 9001 facility

$196.75USD / Each

$196.75USD / Each
Product Description

Key Features

  • With Brij (surfactant)
  • Colorless liquid
  • Density: 1.18
  • Solubility information: miscible
  • pH range: 5.2 to 8.2
  • 4 L natural poly bottle
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

5 Year Warranty
Specifications
  • Concentration25% w/v
  • ClassVolumetric
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)4000
  • PackagingPoly bottle
  • DOT InformationTOXIC LIQUID, INORGANIC, N.O.S. (BARIUM CHLORIDE), 6.1, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN3287
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionBarium Chloride, 25% (w/v) with Surfactant, Volumetric, Natural Poly Bottle, 4 L
  • Warranty5 years
Documents & Certificates
Reviews
Q&A