Reagents Barium Chloride Dihydrate, Purified, Wide-Mouth Container, 500 g

Representative image only.

Reagents Barium Chloride Dihydrate, Purified, Wide-Mouth Container, 500 g

Reagents | Mfr: C2147000-500A5| Item: EW-78910-46

Manufactured using the highest quality raw materials in ISO 9001 facility

$84.00USD / Each

$84.00USD / Each
Product Description

Key Features

  • Colorless/white solid
  • Density: 3.86
  • Solubility information: 370 g/L at 20 °C (68 °F)
  • pH range: 5.2 to 8.2
  • 500 g wide-mouth container
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

10 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaBaCl2·2H2O
  • ClassSalts
  • FormSolid
  • SynonymsBarium chloride 2-hydrate
  • Weight (g)500
  • Formula Weight244.26
  • PackagingWide-mouth container
  • DOT InformationBARIUM COMPOUNDS, N.O.S. (BARIUM CHLORIDE), 6.1, P.G. III
  • Storage SymbolAmbient
  • CAS Number10326-27-9
  • UN NumberUN1564
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Boiling Point C (° C)1560
  • Boiling Point F (° F)2840
  • Melting Point C (° C)963
  • Melting Point F (° F)1765.4
  • DescriptionBarium Chloride Dihydrate, Purified, Wide-Mouth Container, 500 g
  • Warranty10 year
Documents & Certificates
Reviews
Q&A