Reagents Bismuth Nitrate, 0.01M, Standardized, Cubitainer® Package, 10 L

Representative image only.

Reagents Bismuth Nitrate, 0.01M, Standardized, Cubitainer® Package, 10 L

Reagents | Mfr: CB041900-10F| Item: EW-78926-43

Manufactured using the highest quality raw materials in ISO 9001 facility

$250.25USD / Each

$250.25USD / Each
Product Description

Key Features

  • M/100
  • Assay 0.0095 to 0.0105 Molar
  • Colorless liquid
  • Density: 1.1
  • Boiling point: approximately 100 °C (212 °F)
  • Melting point: approximately 0 °C (32 °F)
  • Solubility information: miscible
  • pH range: <1
  • 10 L Cubitainer package
  • 18 month warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

18 Month Warranty
Specifications
  • Concentration 0.01M
  • ClassStandardized
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)10000
  • PackagingCubitainer
  • DOT InformationCORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S. (NITRIC ACID), 8, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN3264
  • Shelf Life (days)548
  • Shelf Life (months)18
  • DescriptionBismuth Nitrate, 0.01M, Standardized, Cubitainer® Package, 10 L
  • Warranty18 months
Documents & Certificates
Reviews
Q&A