Reagents Bromophenol Blue, ACS Reagent, Wide-Mouth Amber Glass Bottle, 5 g

Representative image only.

Reagents Bromophenol Blue, ACS Reagent, Wide-Mouth Amber Glass Bottle, 5 g

Reagents | Mfr: C6106500-5C2| Item: EW-62286-61

Manufactured using the highest quality raw materials in ISO 9001 facility

$46.70USD / Each

$46.70USD / Each
Product Description

Key Features

  • Pale orange solid
  • Density: 0.95
  • Solubility nformation: 4 g/L at 20 °C (68 °F)
  • pH range: 3.0 (yellow) to 4.6 (blue)
  • 5 g wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeACS
  • Linear FormulaC19H10Br4O5S
  • ClassIndicator
  • ChemistryAnalytical Reagents
  • FormSolid
  • Weight (g)5
  • Formula Weight669.96
  • PackagingWide-mouth amber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number115-39-9
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Boiling Point C (° C)279
  • Boiling Point F (° F)534.2
  • Melting Point C (° C)273
  • Melting Point F (° F)523.4
  • DescriptionBromophenol Blue, ACS Reagent, Wide-Mouth Amber Glass Bottle, 5 g
Documents & Certificates
Reviews
Q&A