Reagents Calcium Hydroxide, Purified, Wide-Mouth Container, 500 g

Representative image only.

Reagents Calcium Hydroxide, Purified, Wide-Mouth Container, 500 g

Reagents | Mfr: C2164500-500A5| Item: UX-78910-74

Manufactured using the highest quality raw materials in ISO 9001 facility

$98.30USD / Each

$98.30USD / Each
Product Description

Key Features

  • Minimum Assay 95%
  • Colorless/white solid
  • Density: 2.24
  • Boiling point: 2850 °C (5162 °F) with decomposition
  • Melting point: 580 °C (1076 °F) with decomposition
  • Solubility information: 1.6 g/L at 20 °C (68 °F)
  • 500 g wide-mouth container
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

10 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaCa(OH)2
  • ClassSalts
  • SynonymsSlaked lime, Calcium hydrate
  • FormSolid
  • Weight (g)500
  • Formula Weight74.09
  • Vapor Pressure (torr)< 0.7501
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number1305-62-0
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionCalcium Hydroxide, Purified, Wide-Mouth Container, 500 g
  • Warranty10 year
Documents & Certificates
Reviews
Q&A