Reagents® Calcium Hydroxide, Purified

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Minimum Assay 95%
  • Colorless/white solid
  • Density: 2.24
  • Boiling point: 2850 °C (5162 °F) with decomposition
  • Melting point: 580 °C (1076 °F) with decomposition
  • Solubility information: 1.6 g/L at 20 °C (68 °F)
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$98.30 - $527.00USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Class Weight (g) Description
Availability Pricing
Reagents Calcium Hydroxide, Purified, Fiber Drum, 50 lb EW-62284-77 Mfr # C2164500-50F5
COMPARE
Salts 22680 Calcium Hydroxide, Purified, Fiber Drum, 50 lb
$527.00USD / Each
Reagents Calcium Hydroxide, Purified, Wide-Mouth Container, 500 g EW-78910-74 Mfr # C2164500-500A5
COMPARE
Salts 500 Calcium Hydroxide, Purified, Wide-Mouth Container, 500 g
$98.30USD / Each
Reagents Calcium Hydroxide, Purified, Natural Poly Container, 2.5 kg EW-78919-61 Mfr # C2164500-2.5kg
COMPARE
Salts 2500 Calcium Hydroxide, Purified, Natural Poly Container, 2.5 kg
$339.00USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat