Reagents Chromium Trioxide, 100g, ACS Reagent

Representative image only.

Reagents Chromium Trioxide, 100g, ACS Reagent

Reagents | Mfr: C1179500-100C2| Item: EW-78910-84

Manufactured using the highest quality raw materials in ISO 9001 facility

$117.50USD / Each

$117.50USD / Each
Product Description

Key Features

  • Crystal, Minimum Assay 98%
  • 7 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaCrO3
  • ClassSalts
  • FormSolid
  • Weight (g)100
  • Formula Weight99.99
  • PackagingAmber Jar
  • DOT InformationCHROMIUM TRIOXIDE, ANHYDROUS, 5.1 (6.1, 8)
  • UN NumberUN1463
  • CAS Number1333-82-0
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • DescriptionChromium Trioxide, 100g, ACS Reagent
  • Warranty7 year
Documents & Certificates
Reviews
Q&A