Reagents Congo Red pH Indicator, 0.04% (w/v), Volumetric Grade, 500 mL

Representative image only.

Reagents Congo Red pH Indicator, 0.04% (w/v), Volumetric Grade, 500 mL

Reagents | Mfr: C7108000-500C| Item: EW-78910-96

Manufactured using the highest quality raw materials in ISO 9001 facility

$95.30USD / Each

$95.30USD / Each
Product Description

Key Features

  • pH 3 (Blue) to pH 5 (Red)
  • 0.04% (w/v) Congo Red in 99.96% water
  • Volumetric Grade
  • 2 year warranty

More About this Item

Congo Red is a pH indicator that colors aqueous solutions blue below pH 3.0 and becomes red above pH 5.0. Reagents utilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Chemicals sold by Cole-Parmer® are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Two year warranty
Specifications
  • Purity GradeVolumetric
  • Concentration0.04% w/v
  • Empirical FormulaC₃₂H₂₂N₆Na₂O₆S₂
  • MatrixWater
  • ClassIndicator
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • Formula Weight696.66
  • PackagingAmber Glass Bottle
  • DOT InformationNot Regulated
  • CAS Number573-58-0
  • Shelf Life (days)760
  • Shelf Life (months)24
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • DescriptionCongo Red pH Indicator, 0.04% (w/v), Volumetric Grade, 500 mL
  • Warranty2 year
Documents & Certificates
Reviews
Q&A