Reagents Cupric Sulfate Pentahydrate, ACS Reagent, Wide-Mouth Container, 500g

Representative image only.

Reagents Cupric Sulfate Pentahydrate, ACS Reagent, Wide-Mouth Container, 500g

Reagents | Mfr: C1190500-500A5| Item: EW-78911-03

Manufactured using the highest quality raw materials in ISO 9001 facility

$127.75USD / Each

$127.75USD / Each
Product Description

Key Features

  • Minimum Assay 98%
  • Blue solid
  • Density: 2.29
  • Boiling point: >150 °C (302 °F)
  • Solubility information: 220 g/L at 25 °C (77 °F)
  • 500g wide-mouth container
  • 7 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaCuSO4·5H2O
  • ClassSalts
  • FormSolid
  • SynonymsCupric sulfate 5-hydrate, Copper (II) sulfate pentahydrate
  • Weight (g)500
  • Formula Weight249.69
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7758-99-8
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Melting Point C (° C)110
  • Melting Point F (° F)230
  • DescriptionCupric Sulfate Pentahydrate, ACS Reagent, Wide-Mouth Container, 500g
  • Warranty7 year
Documents & Certificates
Reviews
Q&A