Reagents Deionized Water, ASTM Type IV (D 1193-06), Natural Poly Container, 4 L

Representative image only.

Reagents Deionized Water, ASTM Type IV (D 1193-06), Natural Poly Container, 4 L

Reagents | Mfr: C2346550-4A| Item: UX-78922-48

Manufactured using the highest quality raw materials in ISO 9001 facility

$40.40USD / Each

$40.40USD / Each

Buy It Together

Current Item

Reagents Deionized Water, ASTM Type IV (D 1193-06), Natural Poly Container, 4 L
Reagents Deionized Water, ASTM Type IV (D 1193-06), Natural Poly Container, 4 L
7892248

$40.40USD / Each
+ =
Total Price 57.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Water meets ASTM D 1193 Type IV certification
  • Colorless liquid
  • Density: 1
  • Solubility information: miscible
  • 4 L natural poly container
  • 18 month warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

18 Month Warranty
Specifications
  • Purity GradeASTM Type IV
  • Empirical FormulaH2O
  • ChemistrySolvents
  • SynonymsDihydrogen oxide, Dihydrogen monoxide
  • FormLiquid
  • Volume (mL)4000
  • Formula Weight18.02
  • PackagingPoly container
  • DOT InformationNot regulated by DOT
  • Storage SymbolAmbient
  • CAS Number7732-18-5
  • Shelf Life (days)540
  • Shelf Life (months)18
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • DescriptionDeionized Water, ASTM Type IV (D 1193-06), Natural Poly Container, 4 L
  • Warranty18 months
Documents & Certificates
Reviews
Q&A