Reagents EDTA Disodium Salt, ACS Reagent, Wide-Mouth Container, 500 g

Representative image only.

Reagents EDTA Disodium Salt, ACS Reagent, Wide-Mouth Container, 500 g

Reagents | Mfr: C1201100-500A5| Item: UX-78911-16

Manufactured using the highest quality raw materials in ISO 9001 facility

$243.00USD / Each

$243.00USD / Each
Product Description

Key Features

  • Minimum Assay 99%
  • White solid
  • Density: 0.86
  • Boiling point: >252 °C (485.6 °F), decomposes
  • Solubility information: 110 g/L at 20 °C (68 °F)
  • 500 g wide-mouth container
  • 7 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaC10H14N2O8Na2·2H2O
  • ClassSalts
  • ChemistryOrganics
  • SynonymsEthylenediaminetetraacetic acid, Ethylenedinitrilo)tetraacetic acid, Disodium salt dihydrate
  • FormSolid
  • Weight (g)100
  • Formula Weight372.24
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number6381-92-6
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Melting Point C (° C)252
  • Melting Point F (° F)485.6
  • DescriptionEDTA Disodium Salt, ACS Reagent, Wide-Mouth Container, 500 g
  • Warranty7 year
Documents & Certificates
Reviews
Q&A