Reagents® EDTA Disodium Salt, ACS Reagent

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Minimum Assay 99%
  • White solid
  • Density: 0.86
  • Boiling point: >252 °C (485.6 °F), decomposes
  • Solubility information: 110 g/L at 20 °C (68 °F)
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
$78.80 - $3,640.00USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Packaging Weight (g) Description
Availability Pricing
Reagents EDTA Disodium Salt, ACS Reagent, Poly Pail, 10 lb EW-62281-78 Mfr # C1201100-10F5
COMPARE
Poly pail 4540 EDTA Disodium Salt, ACS Reagent, Poly Pail, 10 lb
$753.00USD / Each
Reagents EDTA Disodium Salt, ACS Reagent, Poly Pail, 10 kg EW-62281-79 Mfr # C1201100-10KG
COMPARE
Poly pail 10000 EDTA Disodium Salt, ACS Reagent, Poly Pail, 10 kg
$1,495.00USD / Each
Reagents EDTA Disodium Salt, ACS Reagent, Wide-Mouth Amber Glass Bottle, 125 g EW-62281-80 Mfr # C1201100-125D2
COMPARE
Wide-mouth amber glass jar 125 EDTA Disodium Salt, ACS Reagent, Wide-Mouth Amber Glass Bottle, 125 g
$78.80USD / Each
Reagents EDTA Disodium Salt, ACS Reagent, Fiber Drum, 50 kg EW-62281-81 Mfr # C1201100-50F4
COMPARE
Fiber drum 50000 EDTA Disodium Salt, ACS Reagent, Fiber Drum, 50 kg
$3,640.00USD / Each
Reagents EDTA Disodium Salt, ACS Reagent, Wide-Mouth Container, 500 g EW-78911-16 Mfr # C1201100-500A5
COMPARE
Wide-mouth container 100 EDTA Disodium Salt, ACS Reagent, Wide-Mouth Container, 500 g
$243.00USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat