Reagents Ethyl Acetate, 4L, Purified

Representative image only.

Reagents Ethyl Acetate, 4L, Purified

Reagents | Mfr: C2199500-4C| Item: EW-78922-83

Manufactured using the highest quality raw materials in ISO 9001 facility

$223.75USD / Each

$223.75USD / Each
Product Description

Key Features

  • Minimum Assay 99%
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

10 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaC4H8O2
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • Volume (mL)4000
  • Formula Weight88.11
  • Vapor Pressure (torr)68.8857
  • PackagingAmber Glass Bottle
  • DOT InformationETHYL ACETATE, 3
  • CAS Number141-78-6
  • UN NumberUN1173
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionEthyl Acetate, 4L, Purified
  • Warranty10 year
Documents & Certificates
Reviews
Q&A