Reagents Ferric Chloride Hexahydrate, Purified, Poly Pail, 12 kg

Representative image only.

Reagents Ferric Chloride Hexahydrate, Purified, Poly Pail, 12 kg

Reagents | Mfr: C2204000-12A5| Item: EW-62285-04

Manufactured using the highest quality raw materials in ISO 9001 facility

$1,130.00USD / Each

$1,130.00USD / Each
Product Description

Key Features

  • Brownish-orange solid
  • Density: 2.9
  • Boiling point: 280 to 285 °C (536 to 545 °F)
  • Solubility information: 912 g/L at 25 °C (77 °F)
  • 12 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaFeCl3
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • SynonymsFerric chloride, Iron (III) chloride, Iron trichloride, Iron(III) chloride hexahydrate
  • Weight (g)12000
  • PackagingPoly pail
  • DOT InformationCORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. (FERRIC CHLORIDE, HEXAHYDRATE), 8, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN3260
  • CAS Number10025-77-1
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)37
  • Melting Point F (° F)98.6
  • DescriptionFerric Chloride Hexahydrate, Purified, Poly Pail, 12 kg
Documents & Certificates
Reviews
Q&A