Reagents Ferric Chloride Hexahydrate, Purified, Wide-Mouth Amber Glass Bottle, 500 g

Representative image only.

Reagents Ferric Chloride Hexahydrate, Purified, Wide-Mouth Amber Glass Bottle, 500 g

Reagents | Mfr: C2204000-500C2| Item: EW-78918-74

Manufactured using the highest quality raw materials in ISO 9001 facility

$87.60USD / Each

$87.60USD / Each
Product Description

Key Features

  • Brownish-orange solid
  • Density: 2.9
  • Boiling point: 280 to 285 °C (536 to 545 °F)
  • Solubility information: 912 g/L at 25 °C (77 °F)
  • 500 g wide-mouth amber glass bottle
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

10 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaFeCl3
  • ClassSalts
  • SynonymsFerric chloride, Iron (III) chloride, Iron trichloride, Iron(III) chloride hexahydrate
  • FormSolid
  • Weight (g)500
  • Formula Weight162.2
  • PackagingWide-mouth amber glass bottle
  • DOT InformationCORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. (FERRIC CHLORIDE, HEXAHYDRATE), 8, P.G. III
  • Storage SymbolAmbient
  • CAS Number10025-77-1
  • UN NumberUN3260
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)37
  • Melting Point F (° F)98.6
  • DescriptionFerric Chloride Hexahydrate, Purified, Wide-Mouth Amber Glass Bottle, 500 g
  • Warranty10 year
Documents & Certificates
Reviews
Q&A