Reagents Ferric Nitrate Nonahydrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 500 g

Representative image only.

Reagents Ferric Nitrate Nonahydrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 500 g

Reagents | Mfr: C1204500-500C2| Item: EW-78918-75

Manufactured using the highest quality raw materials in ISO 9001 facility

$96.50USD / Each

$96.50USD / Each
Product Description

Key Features

  • Minimum Assay 98%
  • Pale mauve solid
  • Density: 1.68
  • Solubility information: 825 g/L at 25 °C (77 °F)
  • 500 g wide-mouth amber glass bottle
  • 3 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

3 Year Warranty
Specifications
  • Purity GradeACS
  • Linear FormulaFe(NO3)3·9H2O
  • ClassSalts
  • SynonymsFerric nitrate, Iron (III) nitrate, Iron(III) nitrate nonahydrate
  • FormSolid
  • Weight (g)500
  • Formula Weight404
  • PackagingWide-mouth amber glass bottle
  • DOT InformationFERRIC NITRATE, 5.1, P.G. III
  • Storage SymbolAmbient
  • CAS Number7782-61-8
  • UN NumberUN1466
  • Shelf Life (days)1125
  • Shelf Life (months)36
  • Boiling Point C (° C)125
  • Boiling Point F (° F)257
  • Melting Point C (° C)47.2
  • Melting Point F (° F)116.96
  • DescriptionFerric Nitrate Nonahydrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 500 g
  • Warranty3 year
Documents & Certificates
Reviews
Q&A