Reagents Formic Acid, 90%, Purified, Natural Poly Container, 500 mL

Representative image only.

Reagents Formic Acid, 90%, Purified, Natural Poly Container, 500 mL

Reagents | Mfr: C2105000-500A| Item: UX-78911-47

Manufactured using the highest quality raw materials in ISO 9001 facility

$75.00USD / Each

$75.00USD / Each
Product Description

Key Features

  • Colorless, liquid
  • Density: 1.22
  • Boiling point: approximately 101 °C (213.8 °F)
  • Flash point: 50 °C (122 °F)
  • Solubility information: miscible
  • pH range: 1.2
  • 500 mL natural poly container
  • 3 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

3 Year Warranty
Specifications
  • Purity GradePurified
  • Concentration0.9
  • Linear FormulaHCOOH
  • ClassAcid
  • FormLiquid
  • Volume (mL)500
  • Formula Weight46.03
  • PackagingPoly container
  • DOT InformationFORMIC ACID, 8 (3), P.G. II
  • Storage SymbolAmbient
  • CAS Number64-18-6
  • UN NumberUN1779
  • Shelf Life (days)1125
  • Shelf Life (months)36
  • DescriptionFormic Acid, 90%, Purified, Natural Poly Container, 500 mL
  • Warranty3 year
Documents & Certificates
Reviews
Q&A