Reagents® Formic Acid, 90%, Purified

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Colorless, liquid
  • Density: 1.22
  • Boiling point: approximately 101 °C (213.8 °F)
  • Flash point: 50 °C (122 °F)
  • Solubility information: miscible
  • pH range: 1.2
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$75.00 - $216.25USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Volume (mL)
Availability Pricing
Reagents Formic Acid, 90%, Purified, Clear Glass Bottle, 4 L EW-62283-46 Mfr # C2105000-4A
COMPARE
Acid Clear glass bottle 4000
$216.25USD / Each
Reagents Formic Acid, 90%, Purified, Natural Poly Container, 500 mL EW-78911-47 Mfr # C2105000-500A
COMPARE
Acid Poly container 500
$75.00USD / Each
Reagents Formic Acid, 90%, Purified, Clear Glass Bottle, 2.5 L EW-78921-24 Mfr # C2105000-2.5D1
COMPARE
Acid Clear glass bottle 2500
$171.50USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat