Reagents Heptanes, Technical, Round Metal Can, 20 L

Representative image only.

Reagents Heptanes, Technical, Round Metal Can, 20 L

Reagents | Mfr: C2213000-20E6| Item: UX-62285-07

Manufactured using the highest quality raw materials in ISO 9001 facility

$438.50USD / Each

$438.50USD / Each
Product Description

Key Features

  • Colorless liquid
  • Density: 0.68
  • Flash point: -4 °C (24.8 °F)
  • Refractive index: 1.385 to 1.389nD
  • Solubility information: 3 µg/mL at 20 °C (68 °F)
  • 20 L round metal can

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeTechnical
  • Linear FormulaC7H16
  • ClassSolvents
  • ChemistrySolvents
  • Synonymsn-Heptane, normal heptane, Dipropyl methane, Heptyl hydride
  • FormLiquid
  • Volume (mL)20000
  • Formula Weight100.2
  • Vapor Pressure (torr)39.7533
  • PackagingRound metal can
  • DOT InformationHEPTANES, 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number142-82-5
  • UN NumberUN1206
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Boiling Point C (° C)98.4
  • Boiling Point F (° F)209.12
  • Melting Point C (° C)-90.6
  • Melting Point F (° F)-131.08
  • DescriptionHeptanes, Technical, Round Metal Can, 20 L
Documents & Certificates
Reviews
Q&A