Reagents® Heptanes, Technical

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Colorless liquid
  • Density: 0.68
  • Flash point: -4 °C (24.8 °F)
  • Refractive index: 1.385 to 1.389nD
  • Solubility information: 3 µg/mL at 20 °C (68 °F)
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$107.25 - $438.50USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Volume (mL)
Availability Pricing
Reagents Heptanes, Technical, Round Metal Can, 20 L EW-62285-07 Mfr # C2213000-20E6
COMPARE
Solvents Round metal can 20000
$438.50USD / Each
Reagents Heptanes, Technical, Amber Glass Bottle, 4 L EW-62285-08 Mfr # C2213000-4C
COMPARE
Solvents Amber glass bottle 4000
$202.75USD / Each
Reagents Heptanes, Technical, Amber Glass Bottle, 1 L EW-78915-58 Mfr # C2213000-1C
COMPARE
Solvents Amber glass bottle 1000
$107.25USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat