Reagents Heptanes, Technical, Amber Glass Bottle, 1 L

Representative image only.

Reagents Heptanes, Technical, Amber Glass Bottle, 1 L

Reagents | Mfr: C2213000-1C| Item: EW-78915-58

Manufactured using the highest quality raw materials in ISO 9001 facility

$107.25USD / Each

$107.25USD / Each
Product Description

Key Features

  • Colorless liquid
  • Density: 0.68
  • Flash point: -4 °C (24.8 °F)
  • Refractive index: 1.385 to 1.389nD
  • Solubility information: 3 µg/mL at 20 °C (68 °F)
  • 1 L amber glass bottle
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

10 Year Warranty
Specifications
  • Purity GradeTechnical
  • Linear FormulaC7H16
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • Synonymsn-Heptane, normal heptane, Dipropyl methane, Heptyl hydride
  • Volume (mL)1000
  • Formula Weight100.2
  • Vapor Pressure (torr)39.7533
  • PackagingAmber glass bottle
  • DOT InformationHEPTANES, 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number142-82-5
  • UN NumberUN1206
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Boiling Point C (° C)98.4
  • Boiling Point F (° F)209.12
  • Melting Point C (° C)-90.6
  • Melting Point F (° F)-131.08
  • DescriptionHeptanes, Technical, Amber Glass Bottle, 1 L
  • Warranty10 year
Documents & Certificates
Reviews
Q&A