Reagents Hydrochloric Acid, 0.10N, Standardized, Cubitainer® Package, 20 L

Representative image only.

Reagents Hydrochloric Acid, 0.10N, Standardized, Cubitainer® Package, 20 L

Reagents | Mfr: C7101000-20F| Item: EW-78927-33

Manufactured using the highest quality raw materials in ISO 9001 facility

$243.50USD / Each

$243.50USD / Each
Product Description

Key Features

  • N/10, Assay 0.0995 to 0.1005 Normal, Aqueous
  • Colorless liquid
  • Density: 1
  • Boiling point: approximately 100 °C (212 °F)
  • Melting point: approximately 0 °C (32 °F)
  • Solubility information: miscible
  • pH range:1
  • 20 L Cubitainer® package
  • 2 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Two year warranty
Specifications
  • Normality0.1
  • ClassStandardized
  • FormLiquid
  • Volume (mL)20000
  • PackagingCubitainer® package
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7647-01-0, 7732-18-5
  • Shelf Life (days)760
  • Shelf Life (months)24
  • DescriptionHydrochloric Acid, 0.10N, Standardized, Cubitainer® Package, 20 L
  • Warranty2 year
Documents & Certificates
Reviews
Q&A