Reagents Hydrochloric Acid, 20° Baumé, Technical, Natural Poly Container, 500 mL

Representative image only.

Reagents Hydrochloric Acid, 20° Baumé, Technical, Natural Poly Container, 500 mL

Reagents | Mfr: C5101000-500A| Item: UX-78911-57

Manufactured using the highest quality raw materials in ISO 9001 facility

$33.20USD / Each

$33.20USD / Each
Product Description

Key Features

  • Colorless to slightly greenish-yellow liquid
  • Density: 1.15
  • Boiling point: approximately 100 °C (212 °F)
  • Melting point: approximately -28 °C (-18.4 °F)
  • Solubility information: miscible
  • pH range: <1
  • 500 mL natural poly container
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

5 Year Warranty
Specifications
  • Purity GradeTechnical
  • Linear FormulaHCl
  • ClassAcid
  • SynonymsMuriatic acid, Hydrogen chloride
  • FormLiquid
  • Volume (mL)500
  • Formula Weight36.46
  • PackagingPoly container
  • DOT InformationHYDROCHLORIC ACID, 8, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1789
  • CAS Number7647-01-0
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionHydrochloric Acid, 20° Baumé, Technical, Natural Poly Container, 500 mL
  • Warranty5 years
Documents & Certificates
Reviews
Q&A