Reagents® Hydrochloric Acid, 20° Baumé, Technical

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Colorless to slightly greenish-yellow liquid
  • Density: 1.15
  • Boiling point: approximately 100 °C (212 °F)
  • Melting point: approximately -28 °C (-18.4 °F)
  • Solubility information: miscible
  • pH range: <1
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$33.20 - $2,485.00USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Description
Availability Pricing
Reagents Hydrochloric Acid, 20° Baumé, Technical, Drum, 15 gal. EW-62286-34 Mfr # C5101000-15E
COMPARE
Acid Drum Hydrochloric Acid, 20° Baumé, Technical, Drum, 15 gal.
$614.00USD / Each
Reagents Hydrochloric Acid, 20° Baumé, Technical, Drum, 200 L EW-62286-35 Mfr # C5101000-200E
COMPARE
Acid Drum Hydrochloric Acid, 20° Baumé, Technical, Drum, 200 L
$2,485.00USD / Each
Reagents Hydrochloric Acid, 20° Baumé, Technical, Natural Poly Container, 500 mL EW-78911-57 Mfr # C5101000-500A
COMPARE
Acid Poly container Hydrochloric Acid, 20° Baumé, Technical, Natural Poly Container, 500 mL
$33.20USD / Each
Reagents Hydrochloric Acid, 20° Baumé, Technical, Natural Poly Container, 1 L EW-78915-79 Mfr # C5101000-1A
COMPARE
Acid Poly container Hydrochloric Acid, 20° Baumé, Technical, Natural Poly Container, 1 L
$51.10USD / Each
Reagents Hydrochloric Acid, 20° Baumé, Technical, Winpak container, 20 L EW-78929-44 Mfr # C5101000-20E7
COMPARE
Acid Winpak Hydrochloric Acid, 20° Baumé, Technical, Winpak container, 20 L
$328.00USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat