Reagents Iodine, ACS Reagent, Wide-Mouth Amber Glass Bottle, 500 g

Representative image only.

Reagents Iodine, ACS Reagent, Wide-Mouth Amber Glass Bottle, 500 g

Reagents | Mfr: C1217500-500C2| Item: EW-78918-77

Manufactured using the highest quality raw materials in ISO 9001 facility

$493.00USD / Each

$493.00USD / Each
Product Description

Key Features

  • Minimum Assay 99.8%
  • Violet-black crystal
  • Density: 4.93
  • Solubility information: 3 g/L at 20 °C (68 °F)
  • Wide-mouth amber glass bottle
  • 500 g
  • 7 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaI2
  • ClassSalts
  • FormSolid
  • Weight (g)500
  • Formula Weight253.81
  • PackagingWide-mouth amber glass bottle
  • DOT InformationIODINE, 8 (6.1), P.G. III
  • Storage SymbolAmbient
  • UN NumberUN3495
  • CAS Number7553-56-2
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Boiling Point C (° C)184
  • Boiling Point F (° F)363.2
  • Melting Point C (° C)114
  • Melting Point F (° F)237.2
  • DescriptionIodine, ACS Reagent, Wide-Mouth Amber Glass Bottle, 500 g
  • Warranty7 year
Documents & Certificates
Reviews
Q&A