Reagents Lactic Acid, 85%, Purified, Natural Poly Container, 500 mL

Representative image only.

Reagents Lactic Acid, 85%, Purified, Natural Poly Container, 500 mL

Reagents | Mfr: C2108000-500A| Item: EW-78911-80

Manufactured using the highest quality raw materials in ISO 9001 facility

$214.25USD / Each

$214.25USD / Each
Product Description

Key Features

  • Colorless liquid
  • Density: 1.21
  • Solubility information: miscible
  • 500 mL natural poly container
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

10 Year Warranty
Specifications
  • Concentration0.85
  • Purity GradePurified
  • Linear FormulaCH3CHOHCOOH
  • ClassAcid
  • FormLiquid
  • Volume (mL)500
  • Viscosity (cSt)36.9
  • Formula Weight90.08
  • Vapor Pressure (torr)0.0813
  • PackagingPoly containerottle
  • DOT InformationCORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S. (LACTIC ACID), 8, P.G. III
  • Storage SymbolAmbient
  • CAS Number50-21-5
  • UN NumberUN3265
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Boiling Point C (° C)122
  • Boiling Point F (° F)251.6
  • Melting Point C (° C)17
  • Melting Point F (° F)62.6
  • DescriptionLactic Acid, 85%, Purified, Natural Poly Container, 500 mL
  • Warranty10 year
Documents & Certificates
Reviews
Q&A