Reagents Magnesium Sulfate Heptahydrate, 500g, Purified

Representative image only.

Reagents Magnesium Sulfate Heptahydrate, 500g, Purified

Reagents | Mfr: C2240000-500A5| Item: EW-78911-90

Manufactured using the highest quality raw materials in ISO 9001 facility

$33.80USD / Each

$33.80USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

10 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaMgSO4·7H2O
  • ClassSalts
  • FormSolid
  • Weight (g)500
  • Formula Weight246.47
  • PackagingPoly Jar
  • CAS Number10034-99-8
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionMagnesium Sulfate Heptahydrate, 500g, Purified
  • Warranty10 year
Documents & Certificates
Reviews
Q&A