Reagents Manganese Sulfate Monohydrate, ACS Reagent, Natural Poly Container, 3 kg

Representative image only.

Reagents Manganese Sulfate Monohydrate, ACS Reagent, Natural Poly Container, 3 kg

Reagents | Mfr: C1243000-3A5| Item: EW-62282-13

Manufactured using the highest quality raw materials in ISO 9001 facility

$692.00USD / Each

$692.00USD / Each
Product Description

Key Features

  • Minimum Assay 98%
  • Pink solid
  • Density: 2.95
  • Boiling point: approximately 850 °C (1562 °F), decomposes
  • Solubility information: 637 g/L at 25 °C (77 °F)
  • 3 kg natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaMnSO4·H2O
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • SynonymsManganese sulfate 1-hydrate
  • Weight (g)3000
  • Formula Weight169.02
  • PackagingPoly container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number10034-96-5
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)700
  • Melting Point F (° F)1292
  • DescriptionManganese Sulfate Monohydrate, ACS Reagent, Natural Poly Container, 3 kg
Documents & Certificates
Reviews
Q&A