Reagents Methanol, ACS Reagent (Absolute), Natural Poly Bottle, 4 L

Representative image only.

Reagents Methanol, ACS Reagent (Absolute), Natural Poly Bottle, 4 L

Reagents | Mfr: C1121000-4A| Item: EW-78919-88

Manufactured using the highest quality raw materials in ISO 9001 facility

$133.50USD / Each

$133.50USD / Each
Product Description

Key Features

  • Minimum Assay 99.8%
  • Colorless liquid
  • Slight, alcohol-like odor
  • Density: 0.79
  • Refractive index: 1.328 to 1.33nD
  • 4 L natural poly bottle
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

5 Year Warranty
Specifications
  • Purity GradeACS
  • Linear FormulaCH3OH
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsMethyl Alcohol, Wood Alcohol
  • Volume (mL)4000
  • Formula Weight32.0419
  • Vapor Pressure (torr)233
  • PackagingPoly bottle
  • DOT InformationMETHANOL, 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number67-56-1
  • UN NumberUN1230
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)64.5
  • Boiling Point F (° F)148.1
  • Melting Point C (° C)-98
  • Melting Point F (° F)-144.4
  • DescriptionMethanol, ACS Reagent (Absolute), Natural Poly Bottle, 4 L
  • Warranty5 years
Documents & Certificates
Reviews
Q&A