Reagents Methanol, Synthetic, Round Metal Can, 20 L

Representative image only.

Reagents Methanol, Synthetic, Round Metal Can, 20 L

Reagents | Mfr: C2121000-20E6| Item: UX-78926-83

Manufactured using the highest quality raw materials in ISO 9001 facility

$297.50USD / Each

$297.50USD / Each
Product Description

Key Features

  • Colorless liquid
  • Density: 0.79
  • Boiling point: approximately 64.5 °C (148.1 °F)
  • Flash point: 11 °C (51.8 °F)
  • Refractive index: 1.328 to 1.33nD
  • Solubility information: miscible
  • 20 L round metal can
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

10 Year Warranty
Specifications
  • Linear FormulaCH3OH
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsMethyl alcohol
  • Volume (mL)20000
  • Formula Weight32.04
  • Vapor Pressure (torr)96.0079
  • PackagingRound metal can
  • DOT InformationMETHANOL, 3, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1230
  • CAS Number67-56-1
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)-98
  • Melting Point F (° F)-144.4
  • DescriptionMethanol, Synthetic, Round Metal Can, 20 L
  • Warranty10 year
Documents & Certificates
Reviews
Q&A