Reagents® Methanol, Synthetic

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Colorless liquid
  • Density: 0.79
  • Boiling point: approximately 64.5 °C (148.1 °F)
  • Flash point: 11 °C (51.8 °F)
  • Refractive index: 1.328 to 1.33nD
  • Solubility information: miscible
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$58.40 - $1,970.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Volume (mL)
Availability Pricing
Reagents Methanol, Synthetic, Drum, 200 L EW-62283-78 Mfr # C2121000-200E
COMPARE
Solvents Drum 200000
$1,970.00USD / Each
Reagents Methanol, Synthetic, Amber Glass Bottle, 1 L EW-78916-09 Mfr # C2121000-1C
COMPARE
Solvents Amber glass bottle 1000
$58.40USD / Each
Reagents Methanol, Synthetic, Natural Poly Container, 4 L EW-78919-87 Mfr # C2121000-4A
COMPARE
Solvents Poly container 4000
$144.25USD / Each
Reagents Methanol, Synthetic, Round Metal Can, 20 L EW-78926-83 Mfr # C2121000-20E6
COMPARE
Solvents Round metal can 20000
$297.50USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat