Reagents n-Pentane, Round Metal Can, 20 L

Representative image only.

Reagents n-Pentane, Round Metal Can, 20 L

Reagents | Mfr: C2265600-20E6| Item: EW-62285-32

Manufactured using the highest quality raw materials in ISO 9001 facility

$722.00USD / Each

$722.00USD / Each
Product Description

Key Features

  • Minimum Assay 98%
  • Colorless liquid
  • Density: 0.63
  • Boiling point: 35 to 36 °C (95 to 96.8 °F)
  • Flash point: -49 °C (-56.2 °F)
  • Solubility information: <1 mg/L at 20 °C (68 °F)
  • 20 L round metal can

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Linear FormulaC5H12
  • ClassSolvents
  • ChemistrySolvents
  • SynonymsPentane
  • FormLiquid
  • Volume (mL)20000
  • Formula Weight72.15
  • Vapor Pressure (torr)385.5317
  • PackagingRound metal can
  • DOT InformationPENTANES, 3, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1265
  • CAS Number109-66-0
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)-130
  • Melting Point F (° F)-202
  • Refractive Index1.35
  • Descriptionn-Pentane, Round Metal Can, 20 L
Documents & Certificates
Reviews
Q&A