Reagents n-Pentane, Safety Coated Amber Glass Bottle, 4 L

Representative image only.

Reagents n-Pentane, Safety Coated Amber Glass Bottle, 4 L

Reagents | Mfr: C2265600-4C1| Item: EW-78920-33

Manufactured using the highest quality raw materials in ISO 9001 facility

$218.00USD / Each

$218.00USD / Each
Product Description

Key Features

  • Minimum Assay 98%
  • Colorless liquid
  • Density: 0.63
  • Boiling point: 35 to 36 °C (95 to 96.8 °F)
  • Flash point: -49 °C (-56.2 °F)
  • Solubility information: <1 mg/L at 20 °C (68 °F)
  • 4 L safety coated amber glass bottle
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

10 Year Warranty
Specifications
  • Linear FormulaC5H12
  • ClassSolvents
  • ChemistrySolvents
  • SynonymsPentane
  • FormLiquid
  • Volume (mL)4000
  • Formula Weight72.15
  • Vapor Pressure (torr)385.5317
  • PackagingSafety coated amber glass bottle
  • DOT InformationPENTANES, 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number109-66-0
  • UN NumberUN1265
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)-130
  • Melting Point F (° F)-202
  • Refractive Index1.35
  • Descriptionn-Pentane, Safety Coated Amber Glass Bottle, 4 L
  • Warranty10 year
Documents & Certificates
Reviews
Q&A