Reagents Nital Etchant, 5% Nitric Acid in Methanol, 2.5L, Volumetric

Representative image only.

Reagents Nital Etchant, 5% Nitric Acid in Methanol, 2.5L, Volumetric

Reagents | Mfr: CN104200-2.5D1| Item: UX-78919-69

Manufactured using the highest quality raw materials in ISO 9001 facility

$178.50USD / Each

$178.50USD / Each
Product Description

Key Features

  • In Methanol
  • 1 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

1 Year Warranty
Specifications
  • ClassBlend
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)2500
  • Vapor Pressure (torr)97
  • PackagingClear Glass Bottle
  • DOT InformationFLAMMABLE LIQUIDS, CORROSIVE, N.O.S. (METHANOL, NITRIC ACID), 3 (8)
  • UN NumberUN2924
  • Shelf Life (days)380
  • Shelf Life (months)12
  • DescriptionNital Etchant, 5% Nitric Acid, 2.5L, Volumetric
  • Warranty1 year
Documents & Certificates
Reviews
Q&A