Reagents® Nital Etchant, Volumetric
Manufactured using the highest quality raw materials in ISO 9001 facility
  • In Methanol
Chemicals sold by Cole-Parmer® are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
$76.60 - $137.25USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Class Volume (mL) Description
Availability Pricing
Reagents Nital Etchant, 5% Nitric Acid in Methanol, 1L, Volumetric EW-78916-17 Mfr # CN104200-1C
COMPARE
Blend 1000 Nital Etchant, 5% Nitric Acid, 1L, Volumetric
$76.60USD / Each
Reagents Nital Etchant, 5% Nitric Acid in Methanol, 2.5L, Volumetric EW-78919-69 Mfr # CN104200-2.5D1
COMPARE
Blend 2500 Nital Etchant, 5% Nitric Acid, 2.5L, Volumetric
$137.25USD / Each