Reagents Phenolphthalein, ACS Reagent, Wide-Mouth Amber Glass Bottle, 25 g

Representative image only.

Reagents Phenolphthalein, ACS Reagent, Wide-Mouth Amber Glass Bottle, 25 g

Reagents | Mfr: C2270000-25C1| Item: UX-62285-37

Manufactured using the highest quality raw materials in ISO 9001 facility

$34.20USD / Each

$34.20USD / Each
Product Description

Key Features

  • White to pale yellow powder
  • Density: 1.3
  • Melting pont: 258 to 262 °C (496.4 to 503.6 °F)
  • Solubility information: 0.4 g/L at 20 °C (68 °F)
  • pH range: 8.0 (colorless) to 10.0 (pink)
  • 25 g wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaC20H14O4
  • ClassIndicator
  • ChemistryAnalytical Reagents
  • FormSolid
  • Weight (g)25
  • Formula Weight318.32
  • PackagingWide-mouth amber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number77-09-8
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionPhenolphthalein, ACS Reagent, Wide-Mouth Amber Glass Bottle, 25 g
Documents & Certificates
Reviews
Q&A