Reagents® Phenolphthalein, ACS Reagent

Manufactured using the highest quality raw materials in ISO 9001 facility
  • White to pale yellow powder
  • Density: 1.3
  • Melting pont: 258 to 262 °C (496.4 to 503.6 °F)
  • Solubility information: 0.4 g/L at 20 °C (68 °F)
  • pH range: 8.0 (colorless) to 10.0 (pink)
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
$34.20 - $1,140.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Class Packaging Weight (g)
Availability Pricing
Reagents Phenolphthalein, ACS Reagent, Wide-Mouth Amber Glass Bottle, 25 g EW-62285-37 Mfr # C2270000-25C1
COMPARE
Indicator Wide-mouth amber glass bottle 25
$34.20USD / Each
Reagents Phenolphthalein, ACS Reagent, Natural Poly Container, 3 kg EW-62285-38 Mfr # C2270000-3KG
COMPARE
Indicator Natural poly container 3000
$1,140.00USD / Each
Reagents Phenolphthalein, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g EW-78912-70 Mfr # C2270000-100C2
COMPARE
Indicator Wide-mouth amber glass bottle 100
$89.40USD / Each
Reagents Phenolphthalein, ACS Reagent, Wide-Mouth Container, 500 g EW-78912-71 Mfr # C2270000-500A5
COMPARE
Indicator Wide-mouth container 500
$277.25USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat