Reagents Phenolphthalein Indicator, 0.25% (w/v) in Methanol, Volumetric, Natural Poly Bottle, 500 mL

Representative image only.

Reagents Phenolphthalein Indicator, 0.25% (w/v) in Methanol, Volumetric, Natural Poly Bottle, 500 mL

Reagents | Mfr: C84003NE-500A| Item: EW-78912-58

Manufactured using the highest quality raw materials in ISO 9001 facility

$59.50USD / Each

$59.50USD / Each
Product Description

Key Features

  • In Methanol
  • Colorless liquid
  • Density: 0.8
  • Flash point: 11 °C (51.8 °F)
  • Solubility information: miscible
  • pH range: 8.0 (colorless) to 10.0 (pink)
  • 500 mL natural poly bottle
  • 2 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Two year warranty
Specifications
  • Concentration0.25% w/v in MeOH
  • ClassIndicator
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly bottle
  • DOT InformationFLAMMABLE LIQUIDS, N.O.S. (METHANOL), 3, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1993
  • Shelf Life (days)730
  • Shelf Life (months)24
  • Melting Point C (° C)-98
  • Melting Point F (° F)-144.4
  • DescriptionPhenolphthalein Indicator, 0.25% (w/v) in Methanol, Volumetric, Natural Poly Bottle, 500 mL
  • Warranty2 year
Documents & Certificates
Reviews
Q&A