Reagents Potash, Sulfurated, 500g

Representative image only.

Reagents Potash, Sulfurated, 500g

Reagents | Mfr: C2275200-500A5| Item: EW-78912-79

Manufactured using the highest quality raw materials in ISO 9001 facility

$141.75USD / Each

$141.75USD / Each
Product Description

Key Features

  • Quality and consistency your lab can count on
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

5 Year Warranty
Specifications
  • Linear FormulaK2S
  • ClassSalts
  • FormSolid
  • Weight (g)100
  • Formula Weight110.26
  • PackagingPoly Jar
  • CAS Number39365-88-3
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionPotash, Sulfurated, 500g,
  • Warranty5 years
Documents & Certificates
Reviews
Q&A