Reagents Potassium Carbonate Anhydrous, 2.5kg, Purified

Representative image only.

Reagents Potassium Carbonate Anhydrous, 2.5kg, Purified

Reagents | Mfr: C2279500-2.5A5| Item: EW-78919-72

Manufactured using the highest quality raw materials in ISO 9001 facility

$162.00USD / Each

$162.00USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • 7 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Linear FormulaK2CO3
  • ClassSalts
  • FormSolid
  • Weight (g)2500
  • Formula Weight138.21
  • PackagingPoly Jar
  • CAS Number584-08-7
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • DescriptionPotassium Carbonate Anhydrous, 2.5kg, Purified
  • Warranty7 year
Documents & Certificates
Reviews
Q&A