Reagents Potassium Chloride, ACS Reagent, Wide-Mouth Amber Glass Bottle, 125 g

Representative image only.

Reagents Potassium Chloride, ACS Reagent, Wide-Mouth Amber Glass Bottle, 125 g

Reagents | Mfr: C1280500-125D2| Item: EW-62282-33

Manufactured using the highest quality raw materials in ISO 9001 facility

$43.30USD / Each

$43.30USD / Each
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless/white solid (granular)
  • Density: 1.98
  • Solubility information: 347 g/L at 20 °C (68 °F)
  • pH range: 5.4 to 8.6
  • 125 g wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaKCl
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)125
  • PackagingWide-mouth amber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7447-40-7
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)1413
  • Boiling Point F (° F)2575.4
  • Melting Point C (° C)771
  • Melting Point F (° F)1419.8
  • DescriptionPotassium Chloride, ACS Reagent, Wide-Mouth Amber Glass Bottle, 125 g
Documents & Certificates
Reviews
Q&A