Reagents Potassium Dichromate, 1.0N, 1L, Standardized

Representative image only.

Reagents Potassium Dichromate, 1.0N, 1L, Standardized

Reagents | Mfr: CP202100-1A| Item: EW-78916-39

Manufactured using the highest quality raw materials in ISO 9001 facility

$68.60USD / Each

$68.60USD / Each
Product Description

Key Features

  • Assay 0.95 to 1.05 Normal, Aqueous
  • 2 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Two year warranty
Specifications
  • Normality1
  • ClassStandard
  • ChemistryStandards
  • FormLiquid
  • Volume (mL)1000
  • PackagingPoly Bottle
  • Shelf Life (days)745
  • Shelf Life (months)24
  • DescriptionPotassium Dichromate, 1.0N, 1L, Standardized
  • Warranty2 year
Documents & Certificates
Reviews
Q&A