Reagents Potassium Ferrocyanide Trihydrate, Purified, Wide-Mouth Amber Glass Bottle, 500 g

Representative image only.

Reagents Potassium Ferrocyanide Trihydrate, Purified, Wide-Mouth Amber Glass Bottle, 500 g

Reagents | Mfr: C2283500-500C2| Item: EW-78918-89

Manufactured using the highest quality raw materials in ISO 9001 facility

$510.00USD / Each

$510.00USD / Each
Product Description

Key Features

  • Yellow granules
  • Density: 1.85
  • Melting point: 70 °C (158 °F), decomposes
  • Solubility information: 360 g/L at 25 °C (77 °F)
  • 500 g wide-mouth amber glass bottle
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

10 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaK4Fe(CN)6·3H2O
  • ClassSalts
  • FormSolid
  • SynonymsPotassium hexacyanoferrate
  • Weight (g)500
  • Formula Weight422.3884
  • PackagingWide-mouth amber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number14459-95-1
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionPotassium Ferrocyanide Trihydrate, Purified, Wide-Mouth Amber Glass Bottle, 500 g
  • Warranty10 year
Documents & Certificates
Reviews
Q&A