Reagents Potassium Fluoride Dihydrate, 500g, Reagent

Representative image only.

Reagents Potassium Fluoride Dihydrate, 500g, Reagent

Reagents | Mfr: C1284000-500A5| Item: EW-78913-01

Manufactured using the highest quality raw materials in ISO 9001 facility

$422.00USD / Each

$422.00USD / Each
Product Description

Key Features

  • Minimum Assay 99%, Crystals
  • 7 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeReagent
  • Linear FormulaKF·2H2O
  • ClassSalts
  • FormSolid
  • Weight (g)100
  • Formula Weight94.13
  • PackagingPoly Jar
  • DOT InformationPOTASSIUM FLUORIDE, SOLID, 6.1
  • CAS Number13455-21-5
  • UN NumberUN1812
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • DescriptionPotassium Fluoride Dihydrate, 500g, Reagent
  • Warranty7 year
Documents & Certificates
Reviews
Q&A