Reagents Potassium Hydroxide, ACS Reagent, Natural Poly Container, 3 kg

Representative image only.

Reagents Potassium Hydroxide, ACS Reagent, Natural Poly Container, 3 kg

Reagents | Mfr: C1285000-3A5| Item: EW-62282-48

Manufactured using the highest quality raw materials in ISO 9001 facility

$337.00USD / Each

$337.00USD / Each
Product Description

Key Features

  • Minimum Assay 85%
  • White deliquescent pellets
  • Density: 2.04
  • Boiling point: approximately 1320 °C (2403 °F)
  • Solubility information: 1210 g/L at 25 °C (77 °F)
  • 3 kg natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaKOH
  • ClassBase
  • ChemistryInorganics
  • FormSolid
  • Weight (g)3000
  • PackagingPoly container
  • DOT InformationPOTASSIUM HYDROXIDE, SOLID, 8, P.G. II
  • Storage SymbolAmbient
  • CAS Number1310-58-3
  • UN NumberUN1813
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • DescriptionPotassium Hydroxide, ACS Reagent, Natural Poly Container, 3 kg
Documents & Certificates
Reviews
Q&A